Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm1
Entrez Gene ID
41852
Gene Name
Tropomyosin 1
Gene Aliases
1305/10, 2299, BcDNA:GH09289, BcDNA:LD37158, BcDNA:SD21996, CG4898, Dm Tm1, Dm TmH33, Dm TmH34, DmTm1, Dmel\CG4898, Dmel_CG4898, PmI, TM, TM1, TMII, Tm, TmH, TmH-33, TmH-34, TmH33, TmH34, TmII, Tmr33, Tmr34, TnH, TnH-33, TnH-34, cTM, cTm, cTmII, chr3R:11122272..11122408, l(3)02299, l(3)S130510, l(3)s2958, mTmII, region 3, tm1, tmII, tropomyosin
Chromosome Location
Chr.3R: 15278220 - 15308779 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003721
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169633.5 NP_732001.3 4 - 5 717 52
NM_169634.3 NP_732002.1 4 - 5 467 52
NM_169635.3 NP_732003.1 4 - 5 900 52
NM_169636.3 NP_732004.1 4 - 5 462 52
NM_169637.4 NP_732005.2 4 - 5 485 52
NM_169638.4 NP_732006.2 6 - 7 1745 52
NM_169639.3 NP_732007.2 4 - 5 1968 52
NM_169640.4 NP_732008.1 2 - 3 1565 52
NM_169641.4 NP_732009.2 3 - 4 1415 52
NM_206495.2 NP_996217.1 4 - 5 485 52
GenBank mRNA AY060262.1 - 2 - 3 2557 52
BT001534.1 - 6 - 7 1743 52
BT001902.1 - 4 - 5 1968 52
BT003786.1 - 3 - 4 1394 52
BT133132.1 - 4 - 5 437 52

View Full Product Details