Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGT8
Entrez Gene ID
374033
Gene Name
UDP glycosyltransferase 8
Gene Aliases
-
Chromosome Location
Chr.4: 56248878 - 56308873 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204204.1 NP_989535.1 4 - 5 1379 115
GenBank mRNA AF129809.1 - 4 - 5 1379 115

View Full Product Details