The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005003.2 | NP_004994.1 | 1 - 2 | 199 | 58 |
GenBank mRNA | AF087660.1 | - | 1 - 2 | 195 | 58 |
AK311877.1 | - | 1 - 2 | 194 | 58 | |
BC058920.1 | - | 1 - 2 | 189 | 58 | |
BI603093.1 | - | 1 - 2 | 238 | 58 | |
EF036489.1 | - | 1 - 2 | 164 | 58 |