The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003222.3 | NP_003213.1 | 5 - 6 | 1167 | 73 |
GenBank mRNA | AK301572.1 | - | 5 - 6 | 983 | 73 |
AK314272.1 | - | 5 - 6 | 1163 | 73 | |
BC035664.1 | - | 5 - 6 | 1126 | 73 | |
BC051829.1 | - | 5 - 6 | 1167 | 73 | |
U85658.1 | - | 5 - 6 | 1090 | 73 | |
X95693.1 | - | 5 - 6 | 970 | 73 |