Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GREB1
Entrez Gene ID
9687
Gene Name
growth regulation by estrogen in breast cancer 1
Gene Aliases
-
Chromosome Location
Chr.2: 11534116 - 11642788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014668.3 NP_055483.2 7 - 8 1196 110
NM_033090.2 NP_149081.1 7 - 8 1147 110
NM_148903.2 NP_683701.2 7 - 8 1212 110
XM_005246192.4 XP_005246249.1 7 - 8 1218 110
XM_011510418.2 XP_011508720.1 6 - 7 1057 110
XM_011510419.2 XP_011508721.1 6 - 7 1057 110
XM_011510423.2 XP_011508725.1 6 - 7 1057 110
GenBank mRNA AF245388.1 - 7 - 8 1196 110
AF245389.1 - 7 - 8 1147 110
AF245390.1 - 7 - 8 1212 110

View Full Product Details