The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130846.3 | NP_001124318.2 | 4 - 5 | 1337 | 75 |
NM_001130847.3 | NP_001124319.1 | 14 - 15 | 1883 | 75 | |
NM_004208.3 | NP_004199.1 | 13 - 14 | 1680 | 75 | |
NM_145812.2 | NP_665811.1 | 13 - 14 | 1668 | 75 | |
NR_132647.1 | 13 - 14 | 1740 | 75 | ||
GenBank mRNA | AF100928.1 | - | 13 - 14 | 1491 | 75 |
AF131759.1 | - | 10 - 11 | 1032 | 75 | |
AK000775.1 | - | 13 - 14 | 1516 | 75 | |
AK314446.1 | - | 13 - 14 | 1468 | 75 | |
AL049703.1 | - | 13 - 14 | 1533 | 75 | |
AL049704.1 | - | 5 - 6 | 612 | 75 | |
BC111065.1 | - | 13 - 14 | 1611 | 75 | |
BC139738.1 | - | 13 - 14 | 1584 | 75 | |
CR457379.1 | - | 13 - 14 | 1449 | 75 | |
DQ016496.2 | - | 4 - 5 | 1337 | 75 | |
DQ016498.1 | - | 14 - 15 | 1660 | 75 | |
DQ016500.1 | - | 13 - 14 | 1517 | 75 |