The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130847.3 | NP_001124319.1 | 8 - 9 | 1089 | 68 |
NM_004208.3 | NP_004199.1 | 8 - 9 | 1089 | 68 | |
NM_145812.2 | NP_665811.1 | 8 - 9 | 1077 | 68 | |
NR_132647.1 | 7 - 8 | 946 | 68 | ||
GenBank mRNA | AF100928.1 | - | 8 - 9 | 900 | 68 |
AF131759.1 | - | 5 - 6 | 441 | 68 | |
AK000775.1 | - | 8 - 9 | 925 | 68 | |
AK314446.1 | - | 8 - 9 | 877 | 68 | |
AL049703.1 | - | 8 - 9 | 942 | 68 | |
AY795556.1 | - | 6 - 7 | 529 | 68 | |
AY795557.1 | - | 5 - 6 | 527 | 68 | |
BC111065.1 | - | 8 - 9 | 1020 | 68 | |
BC139738.1 | - | 8 - 9 | 993 | 68 | |
CR457379.1 | - | 8 - 9 | 858 | 68 | |
DQ016498.1 | - | 8 - 9 | 866 | 68 | |
DQ016500.1 | - | 7 - 8 | 723 | 68 |