The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014668.3 | NP_055483.2 | 13 - 14 | 2253 | 79 |
XM_005246192.4 | XP_005246249.1 | 13 - 14 | 2275 | 79 | |
XM_011510418.2 | XP_011508720.1 | 12 - 13 | 2114 | 79 | |
XM_011510419.2 | XP_011508721.1 | 12 - 13 | 2114 | 79 | |
GenBank mRNA | AF245388.1 | - | 13 - 14 | 2253 | 79 |
BC034455.1 | - | 4 - 5 | 1284 | 79 |