Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GREB1
Entrez Gene ID
9687
Gene Name
growth regulation by estrogen in breast cancer 1
Gene Aliases
-
Chromosome Location
Chr.2: 11534116 - 11642788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014668.3 NP_055483.2 16 - 17 2832 87
XM_005246192.4 XP_005246249.1 16 - 17 2854 87
XM_011510418.2 XP_011508720.1 15 - 16 2693 87
XM_011510419.2 XP_011508721.1 15 - 16 2690 87
GenBank mRNA AF245388.1 - 16 - 17 2832 87
BC034455.1 - 7 - 8 1863 87

View Full Product Details