The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005735.3 | NP_005726.1 | 2 - 3 | 328 | 68 |
XM_017003116.1 | XP_016858605.1 | 2 - 3 | 382 | 68 | |
GenBank mRNA | BC004374.2 | - | 2 - 3 | 196 | 68 |
BC006372.2 | - | 2 - 3 | 161 | 68 | |
BC010090.2 | - | 2 - 3 | 228 | 68 | |
X82207.1 | - | 2 - 3 | 176 | 68 |