The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005499.2 | NP_005490.1 | 5 - 6 | 530 | 79 |
XM_005258404.3 | XP_005258461.2 | 6 - 7 | 811 | 79 | |
XM_006722962.2 | XP_006723025.1 | 4 - 5 | 421 | 79 | |
XM_011526304.2 | XP_011524606.1 | 6 - 7 | 811 | 79 | |
XM_017026134.1 | XP_016881623.1 | 6 - 7 | 811 | 79 | |
GenBank mRNA | AF079566.1 | - | 5 - 6 | 550 | 79 |
AF090384.1 | - | 5 - 6 | 496 | 79 | |
AF110957.1 | - | 5 - 6 | 460 | 79 | |
AK023120.1 | - | 5 - 6 | 531 | 79 | |
AK124730.1 | - | 5 - 6 | 671 | 79 | |
AK299514.1 | - | 2 - 3 | 314 | 79 | |
AL136905.1 | - | 5 - 6 | 485 | 79 | |
BC003153.1 | - | 5 - 6 | 508 | 79 | |
BG503066.1 | - | 5 - 6 | 532 | 79 | |
BT009781.1 | - | 5 - 6 | 460 | 79 | |
CR456756.1 | - | 5 - 6 | 460 | 79 | |
U35832.1 | - | 5 - 6 | 485 | 79 |