The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201466.1 | NP_001188395.1 | 3 - 4 | 439 | 82 |
NM_031916.4 | NP_114122.2 | 2 - 3 | 562 | 82 | |
XM_006714503.2 | XP_006714566.1 | 2 - 3 | 536 | 82 | |
XM_006714504.2 | XP_006714567.1 | 3 - 4 | 438 | 82 | |
XM_017009946.1 | XP_016865435.1 | 3 - 4 | 438 | 82 | |
XM_017009947.1 | XP_016865436.1 | 3 - 4 | 438 | 82 | |
GenBank mRNA | AF239723.1 | - | 2 - 3 | 296 | 82 |
AK098642.1 | - | 2 - 3 | 451 | 82 | |
BC014607.2 | - | 2 - 3 | 523 | 82 | |
BX113042.1 | - | 3 - 4 | 404 | 82 | |
CV024170.1 | - | 2 - 3 | 256 | 82 | |
DB479784.1 | - | 3 - 4 | 439 | 82 |