Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
Chromosome Location
Chr.8: 103298433 - 103332866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164615.1 NP_001158087.1 3 - 4 662 131
NM_001164616.1 NP_001158088.1 4 - 5 574 131
NM_003506.3 NP_003497.2 3 - 4 689 131
NR_133921.1 3 - 4 689 131
GenBank mRNA AB012911.1 - 3 - 4 630 131
AF072873.1 - 3 - 4 652 131
AK299341.1 - 4 - 5 540 131
AK304092.1 - 4 - 5 697 131
AK313789.1 - 3 - 4 633 131
AK316544.1 - 4 - 5 507 131
BC060836.1 - 3 - 4 645 131
BX640609.1 - 3 - 4 656 131

View Full Product Details