The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016248.3 | NP_057332.1 | 11 - 12 | 5580 | 91 |
XM_005266247.1 | XP_005266304.1 | 10 - 11 | 5497 | 91 | |
XM_005266248.3 | XP_005266305.1 | 10 - 11 | 5474 | 91 | |
XM_011534903.2 | XP_011533205.1 | 11 - 12 | 5577 | 91 | |
XM_011534904.1 | XP_011533206.1 | 11 - 12 | 5773 | 91 | |
XM_011534905.1 | XP_011533207.1 | 10 - 11 | 5531 | 91 | |
XM_011534906.2 | XP_011533208.1 | 10 - 11 | 5749 | 91 | |
XM_017020381.1 | XP_016875870.1 | 11 - 12 | 5631 | 91 | |
XM_017020382.1 | XP_016875871.1 | 9 - 10 | 5421 | 91 | |
XM_017020383.1 | XP_016875872.1 | 9 - 10 | 5609 | 91 | |
GenBank mRNA | AB014529.2 | - | 4 - 5 | 3520 | 91 |
AF176555.1 | - | 11 - 12 | 5580 | 91 |