The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016248.3 | NP_057332.1 | 3 - 4 | 227 | 116 |
XM_005266247.1 | XP_005266304.1 | 3 - 4 | 228 | 116 | |
XM_005266248.3 | XP_005266305.1 | 3 - 4 | 256 | 116 | |
XM_005266249.3 | XP_005266306.1 | 3 - 4 | 228 | 116 | |
XM_005266250.3 | XP_005266307.1 | 3 - 4 | 228 | 116 | |
XM_011534903.2 | XP_011533205.1 | 3 - 4 | 224 | 116 | |
XM_011534904.1 | XP_011533206.1 | 3 - 4 | 420 | 116 | |
XM_011534905.1 | XP_011533207.1 | 2 - 3 | 178 | 116 | |
XM_011534906.2 | XP_011533208.1 | 2 - 3 | 396 | 116 | |
XM_017020381.1 | XP_016875870.1 | 3 - 4 | 278 | 116 | |
XM_017020382.1 | XP_016875871.1 | 2 - 3 | 203 | 116 | |
XM_017020383.1 | XP_016875872.1 | 2 - 3 | 391 | 116 | |
GenBank mRNA | AF176555.1 | - | 3 - 4 | 227 | 116 |
CR627087.1 | - | 2 - 3 | 390 | 116 |