Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TES
Entrez Gene ID
26136
Gene Name
testin LIM domain protein
Gene Aliases
TESS, TESS-2
Chromosome Location
Chr.7: 116210493 - 116258786 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015641.3 NP_056456.1 3 - 4 584 77
NM_152829.2 NP_690042.1 3 - 4 418 77
XM_005250258.1 XP_005250315.1 2 - 3 452 77
GenBank mRNA AF245356.1 - 3 - 4 510 77
AF245357.1 - 3 - 4 388 77
AJ250865.1 - 3 - 4 497 77
AK222840.1 - 3 - 4 514 77
AK291802.1 - 3 - 4 399 77
AK315644.1 - 3 - 4 496 77
BC001451.1 - 3 - 4 550 77
GQ423971.1 - 1 - 2 240 77

View Full Product Details