Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tes
- Entrez Gene ID
-
21753
- Gene Name
- testis derived transcript
- Gene Aliases
-
D6Ertd352e, TESS, Tes1, Tes2, testin2
- Chromosome Location
-
Chr.6: 17065149 - 17105825 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC003808.1 |
- |
1 - 2 |
88 |
93 |
View Full Product Details