Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aifm1
Entrez Gene ID
26926
Gene Name
apoptosis-inducing factor, mitochondrion-associated 1
Gene Aliases
AIF, AIFsh2, Hq, Pdcd8
Chromosome Location
Chr.X: 48474944 - 48513563 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290364.1 NP_001277293.1 7 - 8 992 102
NM_012019.3 NP_036149.1 7 - 8 1004 102
GenBank mRNA AF100927.1 - 7 - 8 830 102
AK133764.1 - 7 - 8 844 102
AK180729.1 - 3 - 4 289 101
BC003292.1 - 7 - 8 867 102
BC024882.1 - 7 - 8 1488 102
DQ016499.1 - 7 - 8 867 102

View Full Product Details