Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROPN1L
Entrez Gene ID
697006
Gene Name
rhophilin associated tail protein 1 like
Gene Aliases
-
Chromosome Location
Chr.6: 10408031 - 10433358 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139739.2 XP_014995225.2 - 1635 124
XM_015139740.2 XP_014995226.2 - 1358 124
XM_028849340.1 XP_028705173.1 - 551 124
GenBank mRNA JV595465.1 - - 137 124

View Full Product Details