Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gin1
Entrez Gene ID
316687
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
RGD1307449
Chromosome Location
Chr.9: 111200285 - 111220737 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025012.1 NP_001020183.1 3 - 4 319 100
XM_006245604.1 XP_006245666.1 2 - 3 242 100
XM_006245605.2 XP_006245667.1 3 - 4 738 100
XM_008767377.1 XP_008765599.1 3 - 4 3914 100
XM_008767378.1 XP_008765600.1 3 - 4 3574 100
GenBank mRNA BC082058.1 - 3 - 4 319 100

View Full Product Details