Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tes
Entrez Gene ID
500040
Gene Name
testis derived transcript
Gene Aliases
-
Chromosome Location
Chr.4: 44321883 - 44360407 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039344.2 NP_001034433.1 3 - 4 435 120
GenBank mRNA BC129069.1 - 3 - 4 435 120
DQ339047.1 - 3 - 4 436 120

View Full Product Details