Gene Transcripts
- Gene Symbol
- ARHGEF26-AS1
- Entrez Gene ID
- 100507524
- Gene Name
- ARHGEF26 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 154024401 - 154121210 on Build GRCh38
- UniGene ID
- Hs.370221
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037901.1 |
|
4 |
674 |
|
GenBank mRNA |
BC038431.1 |
- |
4 |
676 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000491862.5 |
- |
4 |
674 |
|
Chr. 3 - 154024401 - 154121210 [-] |
View Full Product Details