Gene Transcripts
- Gene Symbol
- Igkv3-4
- Entrez Gene ID
- 626347
- Gene Name
- immunoglobulin kappa variable 3-4
- Gene Aliases
- Igk-V21-4
- Chromosome Location
- Chr. 6: 70671789 - 70672377 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF045518.1 |
- |
|
221 |
|
BC108385.1 |
- |
|
222 |
|
View Full Product Details