Gene Transcripts

Gene Symbol
SULT1C4
Entrez Gene ID
27233
Gene Name
sulfotransferase family 1C member 4
Gene Aliases
SULT1C, SULT1C2
Chromosome Location
Chr. 2: 108377911 - 108388989 on Build GRCh38
UniGene ID
Hs.312644
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006588.3 NP_006579.2 3 802
XM_017003807.1 XP_016859296.1 3 344
GenBank mRNA BC058861.1 - 2 1299

View Full Product Details