Gene Transcripts

Gene Symbol
Krtap9-1
Entrez Gene ID
16705
Gene Name
keratin associated protein 9-1
Gene Aliases
Chromosome Location
Chr. 11: 99873389 - 99874000 on Build GRCm38
UniGene ID
Mm.422800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015741.2 NP_056556.1 1 545
GenBank mRNA BC116210.2 - 1 494

View Full Product Details