Gene Transcripts
- Gene Symbol
- Ndufa12
- Entrez Gene ID
- 66414
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12
- Gene Aliases
- 2410011G03Rik, AW112974
- Chromosome Location
- Chr. 10: 94199009 - 94220948 on Build GRCm38
- UniGene ID
- Mm.27886
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025551.3 |
NP_079827.2
|
2, 3 |
203 |
|
GenBank mRNA |
AK003131.1 |
- |
2, 3 |
176 |
|
AK003322.1 |
- |
2, 3 |
176 |
|
AK008876.1 |
- |
2, 3 |
176 |
|
AK010460.1 |
- |
2, 3 |
204 |
|
AK032774.1 |
- |
2, 3 |
177 |
|
AK134765.1 |
- |
2, 3 |
176 |
|
AK153240.1 |
- |
2, 3 |
176 |
|
AK167943.1 |
- |
2, 3 |
169 |
|
BC004633.1 |
- |
|
1633 |
|
BC086922.1 |
- |
2, 3 |
208 |
6751147 |
View Full Product Details