Gene Transcripts

Gene Symbol
TIMM22
Entrez Gene ID
29928
Gene Name
translocase of inner mitochondrial membrane 22 homolog (yeast)
Gene Aliases
TEX4, TIM22
Chromosome Location
Chr. 17: 997117 - 1002167 on Build GRCh38
UniGene ID
Hs.745034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013337.2 NP_037469.2 4 712
GenBank mRNA AF155330.1 - 4 691
AK000830.1 - 4 693
AK225453.1 - 4 693
BC002324.2 - 4 712 2819655

View Full Product Details