Gene Transcripts

Gene Symbol
OR10A4
Entrez Gene ID
283297
Gene Name
olfactory receptor family 10 subfamily A member 4
Gene Aliases
JCG5, OR10A4P
Chromosome Location
Chr. 11: 6876625 - 6877619 on Build GRCh38
UniGene ID
Hs.448685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207186.2 NP_997069.2 1 576
GenBank mRNA AF209506.1 - 1 553
BC137024.1 - 1 576 9021019
BC137027.1 - 1 576 9021022

View Full Product Details