Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289032.1 | NP_001275961.1 | 3 | 1079 | |
NM_001289033.1 | NP_001275962.1 | 3 | 924 | ||
NM_006215.3 | NP_006206.2 | 3 | 764 | ||
GenBank mRNA | AK309586.1 | - | 3 | 1057 | |
AK313196.1 | - | 3 | 742 | ||
BC014992.1 | - | 3 | 738 | 4866432 | |
BX248009.1 | - | 3 | 839 | ||
BX248760.1 | - | 3 | 727 | ||
L19684.1 | - | 2 | 672 |