Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030783.1 | NP_110410.1 | 12 | 2407 | |
XM_005253153.1 | XP_005253210.1 | 12 | 2388 | ||
XM_011520391.2 | XP_011518693.1 | 12 | 3025 | ||
XM_011520392.2 | XP_011518694.1 | 13 | 2404 | ||
XM_011520393.2 | XP_011518695.1 | 11 | 2122 | ||
XM_017018368.1 | XP_016873857.1 | 14 | 4799 | ||
XM_017018369.1 | XP_016873858.1 | 13 | 2529 | ||
XM_017018370.1 | XP_016873859.1 | 11 | 2204 | ||
XM_017018371.1 | XP_016873860.1 | 12 | 2318 | ||
GenBank mRNA | AL834357.1 | - | 12 | 2331 | |
BC001210.1 | - | 12 | 2407 | 3503123 |