Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198568.1 | NP_001185497.1 | 5 | 1197 | |
NM_001198592.1 | NP_001185521.1 | 6 | 896 | ||
NM_139247.3 | NP_640340.2 | 6 | 892 | ||
GenBank mRNA | AF497516.1 | - | 5 | 784 | |
AK122714.1 | - | 6 | 892 | ||
AK126468.1 | - | 5 | 746 | ||
BC117473.1 | - | 5 | 873 | 40126024 | |
BC117475.1 | - | 5 | 873 | 40126026 | |
BC143947.1 | - | 5 | 873 | ||
BX248285.1 | - | 6 | 863 | ||
BX537489.1 | - | 1 | 1237 |