Gene Transcripts

Gene Symbol
UBE2R2
Entrez Gene ID
54926
Gene Name
ubiquitin conjugating enzyme E2 R2
Gene Aliases
CDC34B, E2-CDC34B, UBC3B
Chromosome Location
Chr. 9: 33817164 - 33920403 on Build GRCh38
UniGene ID
Hs.643648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017811.3 NP_060281.2 5 1187
XM_005251496.2 XP_005251553.1 4 1120
XM_011517949.1 XP_011516251.1 4 1072
XM_017014860.1 XP_016870349.1 3 985
GenBank mRNA AJ240087.1 - 5 618
AK000426.1 - 5 804
AK225880.1 - 5 792
BC004862.2 - 5 794 3838157
BC047584.1 - 5 1043 4827644
CR457233.1 - 5 613

View Full Product Details