Gene Transcripts

Gene Symbol
PSMA2
Entrez Gene ID
5683
Gene Name
proteasome subunit alpha 2
Gene Aliases
HC3, MU, PMSA2, PSC2
Chromosome Location
Chr. 7: 42916861 - 42932206 on Build GRCh38
UniGene ID
Hs.333786
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002787.4 NP_002778.1 8 671
GenBank mRNA AK222976.1 - 8 639
AK290654.1 - 8 646
AU120930.1 - 8 646
BC002900.2 - 7 597
BC047697.1 - 8 625 5786518
CB990982.1 - 8 714
CR450317.1 - 8 623
D00760.1 - 8 623

View Full Product Details