Gene Transcripts

Gene Symbol
UBXN1
Entrez Gene ID
51035
Gene Name
UBX domain protein 1
Gene Aliases
2B28, SAKS1, UBXD10
Chromosome Location
Chr. 11: 62676498 - 62679117 on Build GRCh38
UniGene ID
Hs.351296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015853.4 NP_056937.2 8 1177
GenBank mRNA BC001372.2 - 8 1115
BC032689.1 - 5 1897

View Full Product Details