Gene Transcripts
- Gene Symbol
- TM2D1
- Entrez Gene ID
- 83941
- Gene Name
- TM2 domain containing 1
- Gene Aliases
- BBP
- Chromosome Location
- Chr. 1: 61681047 - 61725423 on Build GRCh38
- UniGene ID
- Hs.656790
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032027.2 |
NP_114416.1
|
1, 2 |
461 |
|
NR_135160.1 |
|
1, 2 |
461 |
|
NR_135161.1 |
|
1, 2 |
461 |
|
NR_135162.1 |
|
1 |
461 |
|
GenBank mRNA |
AF353990.1 |
- |
1, 2 |
461 |
|
AI168073.1 |
- |
1, 2 |
155 |
|
AK311444.1 |
- |
1, 2 |
107 |
|
AK311705.1 |
- |
1, 2 |
108 |
|
BC029486.1 |
- |
1, 2 |
179 |
5271098 |
BC048995.1 |
- |
1, 2 |
177 |
|
BG700625.1 |
- |
1, 2 |
176 |
|
BU664262.1 |
- |
1 |
105 |
|
View Full Product Details