Gene Transcripts
- Gene Symbol
- FAM90A1
- Entrez Gene ID
- 55138
- Gene Name
- family with sequence similarity 90 member A1
- Gene Aliases
- Chromosome Location
- Chr. 12: 8221260 - 8227618 on Build GRCh38
- UniGene ID
- Hs.196086
- Species
- Homo sapiens
- Gene Symbol
- FAM90A10P
- Entrez Gene ID
- 441328
- Gene Name
- putative protein FAM90A10
- Gene Aliases
- FAM90A10
- Chromosome Location
- Chr. 8: 7769584 - 7771313 on Build GRCh38
- UniGene ID
- Hs.694406
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046351.1 |
|
1 |
132 |
|
GenBank mRNA |
DC395395.1 |
- |
1 |
133 |
|
- Gene Symbol
- FAM90A25P
- Entrez Gene ID
- 389633
- Gene Name
- family with sequence similarity 90, member A7 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 8: 12414521 - 12418032 on Build GRCh38
- UniGene ID
- Hs.650464
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073395.1 |
|
2 |
187 |
|
GenBank mRNA |
AL832996.1 |
- |
2 |
187 |
|
- Gene Symbol
- FAM90A2P
- Entrez Gene ID
- 729689
- Gene Name
- family with sequence similarity 90, member A2 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 8: 12173454 - 12177267 on Build GRCh38
- UniGene ID
- Hs.741074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046354.1 |
|
3 |
333 |
|
GenBank mRNA |
CX762572.1 |
- |
3 |
369 |
|
- Gene Symbol
- FAM90A7P
- Entrez Gene ID
- 441317
- Gene Name
- putative protein FAM90A7
- Gene Aliases
- FAM90A7
- Chromosome Location
- Chr. 8: 7556138 - 7559104 on Build GRCh38
- UniGene ID
- Hs.715411
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046343.1 |
|
1 |
132 |
|
GenBank mRNA |
AK301810.1 |
- |
1 |
133 |
|
View Full Product Details