Gene Transcripts

Gene Symbol
MIER1
Entrez Gene ID
57708
Gene Name
MIER1 transcriptional regulator
Gene Aliases
ER1, MI-ER1
Chromosome Location
Chr. 1: 66924895 - 66988619 on Build GRCh38
UniGene ID
Hs.605432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077700.2 NP_001071168.2 11, 12 1234
NM_001077701.2 NP_001071169.1 10, 11 1090
NM_001077702.2 NP_001071170.2 12, 13 1308
NM_001077703.2 NP_001071171.2 11, 12 1234
NM_001077704.2 NP_001071172.1 10, 11 1090
NM_001146110.1 NP_001139582.1 12, 13 1311
NM_001146111.1 NP_001139583.1 12, 13 1308
NM_001146112.1 NP_001139584.1 12, 13 1269
NM_001146113.1 NP_001139585.1 10, 11 1088
NM_020948.3 NP_065999.2 12, 13 1308
XM_005271076.3 XP_005271133.1 12, 13 1346
XM_011541864.2 XP_011540166.1 13, 14 1253
XM_011541865.2 XP_011540167.1 13, 14 1214
XM_017001919.1 XP_016857408.1 13, 14 1230
XM_017001920.1 XP_016857409.1 13, 14 1247
XM_017001921.1 XP_016857410.1 13, 14 1275
XM_017001922.1 XP_016857411.1 13, 14 1250
XM_017001923.1 XP_016857412.1 11, 12 1070
XM_017001924.1 XP_016857413.1 12, 13 1202
XM_017001925.1 XP_016857414.1 12, 13 1307
XM_017001926.1 XP_016857415.1 13, 14 1246
XM_017001927.1 XP_016857416.1 11, 12 1119
XM_017001928.1 XP_016857417.1 11, 12 1195
XM_017001929.1 XP_016857418.1 12, 13 1176
XM_017001930.1 XP_016857419.1 11, 12 1120
XM_017001931.1 XP_016857420.1 12, 13 1154
XM_017001932.1 XP_016857421.1 10, 11 1083
XM_017001933.1 XP_016857422.1 9, 10 937
XM_017001934.1 XP_016857423.1 8, 9 833
GenBank mRNA AB046830.1 - 8, 9 859
AF515446.1 - 10, 11 1090
AF515447.1 - 11, 12 1171
AF515448.2 - 12, 13 1245
AK302061.1 - 11, 12 1234
AK307717.1 - 12, 13 1205
AL831987.2 - 12, 13 1250
AY124186.2 - 12, 13 1245
AY124187.1 - 11, 12 1171
AY124188.1 - 10, 11 1090
AY124189.2 - 12, 13 1245
AY124190.1 - 11, 12 1171
AY124191.1 - 10, 11 1090
AY124192.2 - 12, 13 1245
AY124193.1 - 11, 12 1171
AY124194.1 - 10, 11 1090
BC108726.1 - 12, 13 1189 5194296
BC125217.1 - 9, 10 886 40123797
BC125218.1 - 9, 10 886 40123803
CR627441.1 - 12, 13 1145

View Full Product Details