Gene Transcripts

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
Chromosome Location
Chr. 3: 157259742 - 157503626 on Build GRCh38
UniGene ID
Hs.658046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167911.1 NP_001161383.1 4, 5 765
NM_001167912.1 NP_001161384.1 4, 5 765
NM_024621.2 NP_078897.2 4, 5 956
XM_011513134.2 XP_011511436.1 3, 4 699
XM_011513135.1 XP_011511437.1 4, 5 595
GenBank mRNA AB051479.1 - 4, 5 765
AK027625.1 - 4, 5 854
AL713656.1 - 3, 4 553
BC101660.1 - 3, 4 592 8069166
BC111017.1 - 4, 5 757 5174889
BC113555.1 - 3, 4 592 8322607

View Full Product Details