Gene Transcripts

Gene Symbol
FASTKD1
Entrez Gene ID
79675
Gene Name
FAST kinase domains 1
Gene Aliases
Chromosome Location
Chr. 2: 169527879 - 169573921 on Build GRCh38
UniGene ID
Hs.529276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281476.2 NP_001268405.1 4, 5 967
NM_001322046.1 NP_001308975.1 4, 5 1640
NM_001322048.1 NP_001308977.1 3, 4 895
NM_001322049.1 NP_001308978.1 3, 4 898
NM_024622.5 NP_078898.3 4, 5 967
NR_104020.2 4, 5 967
XM_006712750.3 XP_006712813.1 3, 4 893
XM_017004911.1 XP_016860400.1 4, 5 2280
GenBank mRNA AB058703.1 - 2, 3 149
AK025554.1 - 1, 2 79
AK055892.1 - 4, 5 878
AK074302.1 - 4, 5 858
AK130797.1 - 4, 5 890
AK225293.1 - 1, 2 79
AL832058.1 - 4, 5 850
BC017950.1 - 3, 4 789
BC032687.2 - 3, 4 296 5581443

View Full Product Details