Gene Transcripts

Gene Symbol
M1AP
Entrez Gene ID
130951
Gene Name
meiosis 1 associated protein
Gene Aliases
C2orf65, D6Mm5e, SPATA37
Chromosome Location
Chr. 2: 74557640 - 74648338 on Build GRCh38
UniGene ID
Hs.348645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281296.1 NP_001268225.1 10 2011
NM_001321739.1 NP_001308668.1 11 2023
NM_138804.4 NP_620159.2 11 2014
XM_005264152.3 XP_005264209.1 6 1171
XM_006711946.3 XP_006712009.1 11 2359
XM_011532548.2 XP_011530850.1 10 2348
XM_011532549.2 XP_011530851.1 11 2301
XM_011532550.2 XP_011530852.1 11 1970
XM_011532551.2 XP_011530853.1 11 1969
XM_011532552.2 XP_011530854.1 10 2186
XM_011532553.2 XP_011530855.1 10 2154
XM_011532554.2 XP_011530856.1 7 1183
GenBank mRNA AK126374.1 - 6 1172
AK300188.1 - 10 2003
BC025997.1 - 11 2017 4823281

View Full Product Details