Gene Transcripts

Gene Symbol
SWSAP1
Entrez Gene ID
126074
Gene Name
SWIM-type zinc finger 7 associated protein 1
Gene Aliases
C19orf39, SWS1AP1, ZSWIM7AP1
Chromosome Location
Chr. 19: 11374707 - 11376951 on Build GRCh38
UniGene ID
Hs.631619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175871.3 NP_787067.2 1 169
GenBank mRNA AK092438.1 - 1 169
BC068071.1 - 1 191 5480023
BC119677.2 - 1 165 40116265

View Full Product Details