Gene Transcripts

Gene Symbol
CDCA5
Entrez Gene ID
113130
Gene Name
cell division cycle associated 5
Gene Aliases
SORORIN
Chromosome Location
Chr. 11: 65061009 - 65084143 on Build GRCh38
UniGene ID
Hs.434886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080668.3 NP_542399.1 6 1534
GenBank mRNA AK093670.1 - 2 1109
AL832526.1 - 5 1250
BC011000.2 - 6 1442 3938081

View Full Product Details