Gene Transcripts

Gene Symbol
ANKRD60
Entrez Gene ID
140731
Gene Name
ankyrin repeat domain 60
Gene Aliases
C20orf86, bA196N14.3
Chromosome Location
Chr. 20: 58216126 - 58228653 on Build GRCh38
UniGene ID
Hs.266571
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304369.1 NP_001291298.1 1, 2 434

View Full Product Details