Gene Transcripts
- Gene Symbol
- VASP
- Entrez Gene ID
- 7408
- Gene Name
- vasodilator-stimulated phosphoprotein
- Gene Aliases
- Chromosome Location
- Chr. 19: 45507430 - 45526989 on Build GRCh38
- UniGene ID
- Hs.515469
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003370.3 |
NP_003361.1
|
6, 7 |
1057 |
|
XM_005259200.1 |
XP_005259257.1
|
6, 7 |
1045 |
|
GenBank mRNA |
AK314812.1 |
- |
6, 7 |
1024 |
|
AL556269.3 |
- |
4, 5 |
530 |
|
BC038224.1 |
- |
6, 7 |
975 |
5210128 |
BG825983.1 |
- |
4, 5 |
608 |
|
KU178381.1 |
- |
5, 6 |
715 |
|
KU178382.1 |
- |
6, 7 |
559 |
|
KU178385.1 |
- |
3, 4 |
264 |
|
Z46389.1 |
- |
6, 7 |
969 |
|
View Full Product Details