Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
64100
Gene Name
epididymal sperm binding protein 1
Gene Aliases
E12, EDDM12, EL149, HE12
Chromosome Location
Chr. 19: 47994651 - 48025153 on Build GRCh38
UniGene ID
Hs.104894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022142.4 NP_071425.3 7 957
XM_017027130.1 XP_016882619.1 7 1056
GenBank mRNA AF279147.1 - 7 957
AJ278478.1 - 7 803
BC015598.2 - 7 921 4649702
GQ149084.1 - 7 921

View Full Product Details