Gene Transcripts

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
Chromosome Location
Chr. 19: 1985438 - 2015703 on Build GRCh38
UniGene ID
Hs.465543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017797.3 NP_060267.2 8 1288
XM_005259593.1 XP_005259650.1 8 1053
XM_011528127.1 XP_011526429.1 6 654
GenBank mRNA AF355797.1 - 8 1288
AK000393.1 - 5 1082
AK027481.1 - 6 650
AK056818.1 - 2 1423
AK225831.1 - 8 1287
AL360275.1 - 5 519
AY007116.1 - 8 1010
BC000564.2 - 5 578
BC008035.2 - 5 572
BC053986.1 - 4 473
BC069199.1 - 6 714
BC094820.1 - 8 1022
CR457230.1 - 4 325

View Full Product Details