Gene Transcripts

Gene Symbol
HEYL
Entrez Gene ID
26508
Gene Name
hes related family bHLH transcription factor with YRPW motif-like
Gene Aliases
HESR3, HEY3, HRT3, bHLHb33
Chromosome Location
Chr. 1: 39623431 - 39639676 on Build GRCh38
UniGene ID
Hs.472566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014571.3 NP_055386.1 5 2219
XM_005270745.2 XP_005270802.1 5 2273
GenBank mRNA BC006087.1 - 5 2189

View Full Product Details