Gene Transcripts

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
Chromosome Location
Chr. 2: 53669980 - 53787009 on Build GRCh38
UniGene ID
Hs.40763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201965.1 NP_001188894.1 8 1629
NM_016115.4 NP_057199.1 9 1463
NM_145863.2 NP_665862.1 8 1254
GenBank mRNA AB209795.1 - 9 1362
AF156778.1 - 8 1261
AK000985.1 - 9 1389
AK001283.1 - 8 1187
AK056069.1 - 8 1631
AK222595.1 - 8 1273
AK222715.1 - 8 1245
AK303133.1 - 7 1146
BC006488.2 - 8 1275 3537359
BC009569.1 - 9 1321 4052577

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
Chromosome Location
Chr. 2: 53670293 - 53860033 on Build GRCh38
UniGene ID
Hs.40763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164165.1 NP_001157637.1 9 1480
GenBank mRNA BC110915.1 - 9 1402

View Full Product Details