Gene Transcripts

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
Chromosome Location
Chr. 9: 116425225 - 117415039 on Build GRCh38
UniGene ID
Hs.601562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184734.1 NP_001171663.1 9 1622
NM_014010.4 NP_054729.3 22 4461
NM_198186.3 NP_937829.3 8 2226
NM_198187.3 NP_937830.3 9 1677
NM_198188.2 NP_937831.1 9 1656
GenBank mRNA AB014534.1 - 22 4461
AF116574.1 - 20 3682
AK024064.1 - 9 1627
AK127350.1 - 21 3682
BC029272.1 - 8 2226
BC146756.1 - 22 4461 9007192
BX649176.1 - 22 3796

View Full Product Details