Gene Transcripts
- Gene Symbol
- TELO2
- Entrez Gene ID
- 9894
- Gene Name
- telomere maintenance 2
- Gene Aliases
- CLK2, TEL2, YHFS
- Chromosome Location
- Chr. 16: 1493344 - 1510459 on Build GRCh38
- UniGene ID
- Hs.271044
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016111.3 |
NP_057195.2
|
11, 12 |
1771 |
|
XM_011522773.2 |
XP_011521075.1
|
11, 12 |
1762 |
|
XM_011522774.2 |
XP_011521076.1
|
11, 12 |
1960 |
|
XM_011522775.2 |
XP_011521077.1
|
11, 12 |
1743 |
|
XM_011522776.2 |
XP_011521078.1
|
11, 12 |
1830 |
|
XM_011522777.2 |
XP_011521079.1
|
11, 12 |
1762 |
|
XM_011522778.2 |
XP_011521080.1
|
11, 12 |
1762 |
|
XM_017023914.1 |
XP_016879403.1
|
11, 12 |
1743 |
|
GenBank mRNA |
AB014583.3 |
- |
11, 12 |
1759 |
|
AK001786.1 |
- |
4, 5 |
480 |
|
AL080126.1 |
- |
11, 12 |
1702 |
|
AL137394.1 |
- |
11, 12 |
1702 |
|
BC017188.2 |
- |
10, 11 |
1527 |
3139743 |
View Full Product Details